Learn More
Sino Biological Recombinant Mouse CTLA-4 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 50503M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CTLA4 (NP_033973.2) (Met 1-Phe 162) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2027