Learn More
Hach Company Sension+ pH gel electrode with ceramic diaphragm (general use)
Catalog No. 501997968 Shop All Hach Company Products
Encompass
Low cost gel-filled pH electrode with ceramic diaphragm. Automatic temperature compensation. Polycarbonate body. Range 0-14 pH, 0-80 °C. 1 m cable. For portable meters.This low maintenance pH electrode is suitable for measurements in aqueous media in general. A storage protector prevents breakage and avoids over pressurisation of the diaphragm.
Catalog No. 50-199-7968 Supplier Hach Company Supplier No. LZW5050T.97.002
Low cost gel-filled pH electrode with ceramic diaphragm. Automatic temperature compensation. Polycarbonate body. Range 0-14 pH, 0-80 °C. 1 m cable. For portable meters.This low maintenance pH electrode is suitable for measurements in aqueous media in general. A storage protector prevents breakage and avoids over pressurisation of the diaphragm.