Learn More
Sino Biological Recombinant Mouse CD19 Protein (ECD, His Tag)
Supplier:  Sino Biological 50510M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD19 (NP_033974.2) (Met1-Gly287) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2036