Learn More
Sino Biological Recombinant Mouse LRIG1 Protein (His Tag)
Supplier:  Sino Biological 50511M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse LRIG1 (NP_032403.2) (Met 1-Thr 794) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2038