Learn More
Sino Biological Recombinant Mouse Mer Protein (His & GST Tag)
Supplier:  Sino Biological 50514M20B150
Encompass_Preferred
A DNA sequence encoding the mouse MERTK (Q60805) (Glu573-Tyr867) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2042