Learn More
Hach Company Sension+ 5051T portable combination pH electrode for "dirty" (wastewater) applications
Catalog No. 501997969 Shop All Hach Company Products
Encompass
Sension+ 5051T is a combination pH electrode with a glass body (protected by a plastic shroud), non-refillable solid-electrolyte reference and built-in temperature sensor. The 5051T has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable pH meters. The 5051T has an open annular junction and is ideal for pH measurements in wastewater, foods, and other “dirty” samples with high solids content.
Catalog No. 50-199-7969 Supplier Hach Company Supplier No. LZW5051T.97.002
Sension+ 5051T is a combination pH electrode with a glass body (protected by a plastic shroud), non-refillable solid-electrolyte reference and built-in temperature sensor. The 5051T has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable pH meters. The 5051T has an open annular junction and is ideal for pH measurements in wastewater, foods, and other “dirty” samples with high solids content.