Learn More
Sino Biological Recombinant Mouse DLL1 Protein (His Tag)
Supplier:  Sino Biological 50522M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse DLL1 (NP_031891.2) (Met 1-Gln 516) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2047