Learn More
World Precision Instrument WPI Small Bird Adaptor Kit
SDP
Catalog No. 502539174
Encompass
  • The Small Bird Adaptor includes the 45° ear bars (nontraumatic) required for birds.
  • The Small Bird Adaptor uses a beak support and “V” shaped beak clamp to hold the beak down and centered, in place of the incisor bar/nose clamp arrangement used for rats
Catalog No. 50-253-9174 Supplier World Precision Instrument Supplier No. 505241
  • The Small Bird Adaptor includes the 45° ear bars (nontraumatic) required for birds.
  • The Small Bird Adaptor uses a beak support and “V” shaped beak clamp to hold the beak down and centered, in place of the incisor bar/nose clamp arrangement used for rats