Learn More
Sino Biological Recombinant Mouse CD99L2 Protein (His Tag)
Supplier:  Sino Biological 50526M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD99L2 (NP_612182.1) extracellular domain (Met 1-Ala 164) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2057