Learn More
Hach Company Sension+ 5052T portable combination pH electrode for "difficult" (LIS, high temperature) applications
Catalog No. 501997970 Shop All Hach Company Products
Encompass
Sension+ 5052T is a combination pH electrode with a glass body (protected by a plastic shroud), non-refillable gel reference electrolyte and built-in temperature sensor. The 5052T has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable pH meters. The 5052T has a large porous annular PTFE junction and is ideal for pH measurements in low ionic strength, high temperature, viscous, or other “difficult” sample types.
Catalog No. 50-199-7970 Supplier Hach Company Supplier No. LZW5052T.97.002
Sension+ 5052T is a combination pH electrode with a glass body (protected by a plastic shroud), non-refillable gel reference electrolyte and built-in temperature sensor. The 5052T has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable pH meters. The 5052T has a large porous annular PTFE junction and is ideal for pH measurements in low ionic strength, high temperature, viscous, or other “difficult” sample types.