Learn More
Sino Biological Recombinant Mouse CSRP1 Protein (His Tag)
Supplier:  Sino Biological 50532M08E100
Encompass_Preferred
A DNA sequence encoding the mouse CSRP1 (P97315) (Met 1-Glu 193) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-162-2067