Learn More
Sino Biological Recombinant Mouse CD2 Protein (His Tag)
Supplier:  Sino Biological 50537M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD2 (NP_038514.1) (Met 1-Ser 203) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2072