Learn More
Sino Biological Recombinant Mouse CADM1 Protein (His Tag)
Supplier:  Sino Biological 50538M08H50
Encompass_Preferred
A DNA sequence encoding the mouse Cadm1 (NP_061240.3) (Met1-His377) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2074