Learn More
Sino Biological Recombinant Mouse Factor D Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50539M08H50
Encompass_Preferred
A DNA sequence encoding the full length of mouse CFD isoform 2 (P03953-2) (Met 1-Ser 258) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2076