Learn More
Sino Biological Recombinant Mouse TGF-beta RIII/TGFBR3 Protein (His Tag)
Supplier:  Sino Biological 50542M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TGFBR3 (NP_035708.2) (Met1-Thr785) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2081