Learn More
Sino Biological Recombinant Mouse C4.4A/LYPD3 Protein (His Tag)
Supplier:  Sino Biological 50546M08H100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment of mouse LYPD3 (NP_598504.1) (Met 1-His 287) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2089