Learn More
Sino Biological Recombinant Mouse Peroxiredoxin 1 Protein (His Tag)
Supplier:  Sino Biological 50552M08E50
Encompass_Preferred
A DNA sequence encoding the mouse PRDX1 (P35700) (Met 1-Lys 199) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2094