Learn More
Sino Biological Recombinant Mouse Frizzled-10 Protein (His Tag)
Supplier:  Sino Biological 50568M08H100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment of mouse FZD10 (NP_780493.1) (Met 1-Gly 162) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2113