Learn More
Sino Biological Recombinant Mouse CDNF Protein (His Tag)
Supplier:  Sino Biological 50569M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CDNF (NP_808315.1) (Met 1-Leu 187) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2115