Learn More
Hach Company Sension+ 5057 laboratory ORP electrode for general applications
Catalog No. 501997973 Shop All Hach Company Products
Encompass
Sension+ 5057 is a combination platinum ORP electrode with a glass body and refillable liquid-electrolyte reference. The 5057 has a fixed 1 meter cable and BNC connector dedicated for use with Hach Sension+ Laboratory pH (mV) meters. The 5057 has a ceramic junction is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.
Catalog No. 50-199-7973 Supplier Hach Company Supplier No. LZW5057.97.0002
Sension+ 5057 is a combination platinum ORP electrode with a glass body and refillable liquid-electrolyte reference. The 5057 has a fixed 1 meter cable and BNC connector dedicated for use with Hach Sension+ Laboratory pH (mV) meters. The 5057 has a ceramic junction is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.