Learn More
Sino Biological Recombinant Mouse Contactin-4 Protein (His Tag)
Supplier:  Sino Biological 50570M08B50
Encompass_Preferred
A DNA sequence encoding the mouse Cntn4 (Met1-Gly999) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2118