Learn More
Sino Biological Recombinant Mouse CEACAM-1/CD66a Protein (ECD, hFc Tag)
Supplier:  Sino Biological 50571M02H200
Encompass_Preferred
A DNA sequence encoding the mouse Ceacam1 (NP_001034274.1) (Met1-Gly428) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2119