Learn More
Sino Biological Recombinant Mouse EphA4 Protein (His Tag)
Supplier:  Sino Biological 50575M08H200
Encompass_Preferred
A DNA sequence encoding the mouse EPHA4 (NP_031962.2) extracellular domain (Met 1-Thr 547) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2130