Learn More
Sino Biological Recombinant Mouse 14-3-3 sigma Protein
Supplier:  Sino Biological 50576MNCE100
Encompass_Preferred
A DNA sequence encoding the mouse SFN (NP_061224.2) (Met 1-Ser 248) was expressed and purified, with additional two amino acids (Gly & Pro) at the N-terminu.
Catalog No. 50-162-2131