Learn More
Sino Biological Recombinant Mouse Ephrin A2/EFNA2 Protein (His Tag)
Supplier:  Sino Biological 50579M08H200
Encompass_Preferred
A DNA sequence encoding the mouse EFNA2 (NP_031935.3) without the pro peptide (Met 1-Asn 184) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2136