Learn More
Sino Biological Recombinant Mouse Ephrin-B1 Protein (hFc Tag)
Supplier:  Sino Biological 50580M02H100
Encompass_Preferred
A DNA sequence encoding the mouse EFNB1 (NP_034240.1) extracellular domain (Met 1-Ser 229) was as fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2140