Learn More
Sino Biological Recombinant Mouse Vitronectin Protein (His Tag)
Supplier:  Sino Biological 50585M08H100
Encompass_Preferred
A DNA sequence encoding the mouse vitronectin (NP_035837.1) (Met 1-Lys 478) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2158