Learn More
Sino Biological Recombinant Mouse EphA7 Protein (His Tag)
Supplier:  Sino Biological 50587M08H200
Encompass_Preferred
A DNA sequence encoding the mouse EPHA7 isoform 1 (Q61772-1) extracellular domain (Met 1-Ile 556) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2164