Learn More
Sino Biological Recombinant Mouse beta-Catenin Protein (His & GST Tag)
Supplier:  Sino Biological 50592M20B20
Encompass_Preferred
A DNA sequence encoding the mouse CTNNB1 (Q02248) (Met1-Leu781) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2170