Learn More
Sino Biological Recombinant Mouse Ephrin-A1 Protein (hFc Tag)
Supplier:  Sino Biological 50593M02H100
Encompass_Preferred
A DNA sequence encoding the mouse EFNA1 (NP_034237.3) without the pro peptide (Met 1-Ser 182) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2171