Learn More
Sino Biological Recombinant Mouse Ephrin A3 Protein (His Tag)
Supplier:  Sino Biological 50594M08H100
Encompass_Preferred
A DNA sequence encoding the mouse EFNA3 (NP_034238.1) without the pro peptide (Met 1-Ser 205) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2176