Learn More
Sino Biological Recombinant Mouse Ephrin A5 Protein (hFc Tag)
Supplier:  Sino Biological 50597M02H100
Encompass_Preferred
A DNA sequence encoding the mouse EFNA5 isoform long (O08543-1) without the pro peptide (Met 1-Asn 203) was as fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2185