Learn More
Sino Biological Recombinant Mouse Epiregulin Protein (hFc Tag)
Supplier:  Sino Biological 50599M01H100
Encompass_Preferred
A DNA sequence encoding the mature form of mouse EREG (Q61521) (Val 56-Leu 101) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-2195