Learn More
Waters Corp WATERS CORPORATION Mixer Assembly, 50 μL, for 1525μ Series Pumps
Catalog No. 50792735 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 50 μL Mixer Assembly is a replacement component designed specifically for Waters 1525μ series pumps, ensuring optimal performance and fluidic integrity. It serves as a crucial spare part for maintaining and repairing your Waters chromatography system.
  • Has a 50 μL mixer volume.
  • Designed for Waters 1525μ series pumps.
  • Serves as a spare part for maintenance and repair.
Catalog No. 50-792-735 Supplier Waters Corp Supplier No. 700002306
This 50 μL Mixer Assembly is a replacement component designed specifically for Waters 1525μ series pumps, ensuring optimal performance and fluidic integrity. It serves as a crucial spare part for maintaining and repairing your Waters chromatography system.
  • Has a 50 μL mixer volume.
  • Designed for Waters 1525μ series pumps.
  • Serves as a spare part for maintenance and repair.