Learn More
Sino Biological Recombinant Mouse NEGR1 Protein (His Tag)
Supplier:  Sino Biological 50601M08H100
Encompass_Preferred
A DNA sequence encoding the mouse NEGR1 (NP_001034183.1) (Met 1-Gly 318) without the pro peptide was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2198