Learn More
Sino Biological Recombinant Mouse ACVR2A Protein (His & hFc Tag)
Supplier:  Sino Biological 50613M03H200
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse ACVR2A (NP_031422.3) (Met 1-Pro 134) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2205