Learn More
Sino Biological Recombinant Mouse ACVR2A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50613M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse ACVR2A (NP_031422.3) (Met 1-Pro 134) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2206