Learn More
Hach Company Sension+ 5062 portable titanium conductivity cell for “difficult” applications
Catalog No. 501997976 Shop All Hach Company Products
Encompass
Sension+ 5062 is a two-pole titanium conductivity cell with a titanium body and built-in temperature sensor. The 5062 has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable Conductivity meters. The 5062 is ideal for conductivity measurements in extreme conditions and for “difficult” applications, including dirty and viscous samples.
Catalog No. 50-199-7976 Supplier Hach Company Supplier No. LZW5062.97.0002
Sension+ 5062 is a two-pole titanium conductivity cell with a titanium body and built-in temperature sensor. The 5062 has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable Conductivity meters. The 5062 is ideal for conductivity measurements in extreme conditions and for “difficult” applications, including dirty and viscous samples.