Learn More
Sino Biological Recombinant Mouse SLAMF8 Protein (His Tag)
Supplier:  Sino Biological 50623M08H100
Encompass_Preferred
A DNA sequence encoding the mouse SLAMF8 (NP_083360.2) extracellular domain (Met 1-Asp 231) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2216