Learn More
Sino Biological Recombinant Mouse PLK1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50624M07B50
Encompass_Preferred
A DNA sequence encoding the mouse PLK1 (Q07832) (Met 1-Ser 603) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2219