Learn More
Sino Biological Recombinant Mouse PGK1 Protein (His Tag)
Supplier:  Sino Biological 50628M08B50
Encompass_Preferred
A DNA sequence encoding the mouse PGK1 (P09411) (Met 1-Val 417) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2228