Learn More
Sino Biological Recombinant Mouse Semaphorin 3A Protein (hFc Tag)
Supplier:  Sino Biological 50631M01H50
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Lys 26-Phe 546) of mouse SEMA3A (O08665) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-2234