Learn More
Sino Biological Recombinant Mouse CD200R4 Protein (hFc Tag)
Supplier:  Sino Biological 50639M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD200R4 (NP_997127.1) (Met 1-Thr 241) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2241