Learn More
Sino Biological Recombinant Mouse DLL4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50640M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse DLL4 (NP_062327.2) (Met 1-Pro 525) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2245