Learn More
Sino Biological Recombinant Mouse Semaphorin 4D Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50642M08H100
Encompass_Preferred
A DNA sequence encoding the mouse SEMA4D (NP_038688.2) extracellular domain (Met 1-Arg 733) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2249