Learn More
Sino Biological Recombinant Mouse ICAM-2/CD102 Protein (hFc Tag)
Supplier:  Sino Biological 50644M02H100
Encompass_Preferred
A DNA sequence encoding the mouse ICAM2 (NP_034624.1) extracellular domain (Met 1-Gln 222) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2253