Learn More
Sino Biological Recombinant Mouse ICAM-2/CD102 Protein
Supplier:  Sino Biological 50644MCCH100
Encompass_Preferred
A DNA sequence encoding the mouse ICAM2 (NP_034624.1) (Met 1-Gln 222)was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-162-2255