Learn More
Sino Biological Recombinant Mouse FABP4/A-FABP Protein (His Tag)
Supplier:  Sino Biological 50652M08H100
Encompass_Preferred
A DNA sequence encoding the mouse FABP4 (NP_077717.1) (Met 1-Ala 132) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2265