Learn More
Sino Biological Recombinant Mouse GAD65/GAD2 Protein (His & GST Tag)
Supplier:  Sino Biological 50653M20B50
Encompass_Preferred
A DNA sequence encoding full length of mouse GAD2 (NP_032104.2) (Met 1-Leu 585) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2270