Learn More
Sino Biological Recombinant Mouse GAD65/GAD2 Protein
Supplier:  Sino Biological 50653MNCB20
Encompass_Preferred
A DNA sequence encoding the mouse GAD2 (NP_032104.2) (Met 1-Leu 585) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-162-2272